Learn More
Medchemexpress LLC L-Carnosine (Standard) | 305-84-0 | 99.8% | 226.23 | 100 MG
SDP
Catalog No. 5000454311
Encompass_Preferred
L-Carnosine (Standard) is the analytical standard of L-Carnosine, intended for research and analytical applications. L-Carnosine is a dipeptide of the amino acids beta-alanine and histidine, with the potential to suppress many of the biochemical changes that accompany aging.
  • Is a dipeptide of the amino acids beta-alanine and histidine and has the potential to suppress many of the biochemical changes that accompany aging.
  • Also exhibits some antioxidant effects.
  • The antioxidant mechanism is attributed to its chelating effect against metal ions, superoxide dismutase (SOD)-like activity, and ROS and free radicals scavenging ability.
Catalog No. 50-004-54311 Supplier Medchemexpress LLC Supplier No. HYW013494R100MG
L-Carnosine (Standard) is the analytical standard of L-Carnosine, intended for research and analytical applications. L-Carnosine is a dipeptide of the amino acids beta-alanine and histidine, with the potential to suppress many of the biochemical changes that accompany aging.
  • Is a dipeptide of the amino acids beta-alanine and histidine and has the potential to suppress many of the biochemical changes that accompany aging.
  • Also exhibits some antioxidant effects.
  • The antioxidant mechanism is attributed to its chelating effect against metal ions, superoxide dismutase (SOD)-like activity, and ROS and free radicals scavenging ability.