Learn More
Cayman Chemical L-Carntne 25g
SDP
Catalog No. 502995655
Encompass_Preferred
A conditionally essential nutrient; facilitates the transport of long-chain fatty acids into the mitochondrial matrix for β-oxidation, has other diverse roles on metabolism, and is involved in the maintenance of CoA stores; plasma and/or tissue levels are decreased in primary L-carnitine deficiency; serum and tissue levels are also reduced in secondary L-carnitine deficiencies
Catalog No. 50-299-5655 Supplier Cayman Chemical Supplier No. 2148925 G
A conditionally essential nutrient; facilitates the transport of long-chain fatty acids into the mitochondrial matrix for β-oxidation, has other diverse roles on metabolism, and is involved in the maintenance of CoA stores; plasma and/or tissue levels are decreased in primary L-carnitine deficiency; serum and tissue levels are also reduced in secondary L-carnitine deficiencies