Learn More
ATCC L Cells; Fibroblast; Mouse (Mus musculus)
Catalog No. 502384860
Encompass
L Cells is a fibroblast cell line that was isolated from the areolar adipose tissue of a 100-day-old, male mouse. It is used to obtain a control conditioned medium for comparison to Wnt-3A or Wnt-5A conditioned medium from L Wnt-3A cells or L Wnt-5A cells. This cell line was deposited by R Nusse.
Catalog No. 50-238-4860 Supplier ATCC Supplier No. CRL2648
 Restricted Item
Add to Cart
Add to Cart
L Cells is a fibroblast cell line that was isolated from the areolar adipose tissue of a 100-day-old, male mouse. It is used to obtain a control conditioned medium for comparison to Wnt-3A or Wnt-5A conditioned medium from L Wnt-3A cells or L Wnt-5A cells. This cell line was deposited by R Nusse.