Learn More
Medchemexpress LLC L-Chicoric Acid | 70831-56-0 | 474.37 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0457A5MG
Encompass_Preferred
L-Chicoric Acid is a dicaffeoyltartaric acid that acts as a potent, selective, and reversible HIV-1 integrase inhibitor with an IC50 of approximately 100 nM. It has demonstrated the ability to inhibit HIV-1 replication in tissue culture, affecting viral entry and integration processes.
  • Potent, selective, and reversible HIV-1 integrase inhibitor
  • Inhibits HIV-1 replication in tissue culture
  • Purity of 99.85% and ee. of 99.65%
  • Classified as a phenylpropanoid, simple phenylpropanol, phenol, and polyphenol
  • Inhibits integration at concentrations from 500 nM to 10 μM
  • Affects viral entry at concentrations of 5 μM and higher
  • For research use only
Catalog No. 50-003-89014