Learn More
Abcam L-cis-Diltiazem, cyclic nucleotide-gated channel blocker, 5MG
Catalog No. 502707516
Encompass_Preferred
MW 451 Da, Purity >97%. Selective inhibitor of cyclic nucleotide-gated (CNG) channels. Blocks rod-type CNGA1, olfactory-type CNGA2 and cone-type CNGA3, at micromolar concentrations. Optical isomer of diltiazem (ab120260). Protects against myocardial dysfunction in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7516 Supplier Abcam Supplier No. AB1205325MG
MW 451 Da, Purity >97%. Selective inhibitor of cyclic nucleotide-gated (CNG) channels. Blocks rod-type CNGA1, olfactory-type CNGA2 and cone-type CNGA3, at micromolar concentrations. Optical isomer of diltiazem (ab120260). Protects against myocardial dysfunction in vitro.
The product is subject to the following: Abcam Restricted Use Statement