Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-citronellol | 7540-51-4 | MFCD00063214 | 99.5% | 156.27 | C10H20O | 100 G
SDP

Catalog No. 5000302961
Encompass_Preferred

L-citronellol is an analytical standard of the monoterpenoid alcohol (S)-3,7-dimethyloct-6-en-1-ol supplied for research and analytical applications. The material is provided with supporting documentation (COA, SDS) and should be stored under the conditions specified in the certificate of analysis. For research use only.

  • Purity 99.5%.
  • Molecular formula C10H20O.
  • Molecular weight 156.27 g/mol.
  • CAS number 7540-51-4.
  • Intended for research and analytical applications.
  • Supplied with certificate of analysis and safety data sheet.
  • Available sizes listed by manufacturer up to 5 g (see COA for batch packaging).

Catalog No. 50-003-02961 Supplier Medchemexpress LLC Supplier No. HYW015515100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-citronellol is an analytical standard of the monoterpenoid alcohol (S)-3,7-dimethyloct-6-en-1-ol supplied for research and analytical applications. The material is provided with supporting documentation (COA, SDS) and should be stored under the conditions specified in the certificate of analysis. For research use only.

  • Purity 99.5%.
  • Molecular formula C10H20O.
  • Molecular weight 156.27 g/mol.
  • CAS number 7540-51-4.
  • Intended for research and analytical applications.
  • Supplied with certificate of analysis and safety data sheet.
  • Available sizes listed by manufacturer up to 5 g (see COA for batch packaging).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.