Learn More
Medchemexpress LLC HY-N0391 1g Medchemexpress, L-Citrulline CAS:372-75-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN03911G
Encompass_Preferred
Medchemexpress, HY-N0391 1g L-Citrulline CAS:372-75-8 L-Citrulline is an amino acid derived from ornithine in the catabolism of proline or glutamine and glutamate, or from l-arginine via arginine-citrulline pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3553