Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Citrulline | 372-75-8 | 175.19 | 100 MG
SDP

Catalog No. 5000389135
Encompass_Preferred

L-Citrulline (Standard) is the analytical standard of L-Citrulline, intended for research and analytical applications. It is an amino acid derived from ornithine in the catabolism of proline or glutamine and glutamate, or from l-arginine via the arginine-citrulline pathway. The compound is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS as an analytical standard.

  • Purity: 99.89%
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Amino acids
  • Initial source: Endogenous metabolite, disease markers, endocrine diseases, nervous system disorder
  • Application: Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS as an analytical standard.

Catalog No. 50-003-89135 Supplier Medchemexpress LLC Supplier No. HYN0391R100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Citrulline (Standard) is the analytical standard of L-Citrulline, intended for research and analytical applications. It is an amino acid derived from ornithine in the catabolism of proline or glutamine and glutamate, or from l-arginine via the arginine-citrulline pathway. The compound is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS as an analytical standard.

  • Purity: 99.89%
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Amino acids
  • Initial source: Endogenous metabolite, disease markers, endocrine diseases, nervous system disorder
  • Application: Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS as an analytical standard.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.