Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Citrulline, 99.3%, MP Biomedicals™

Catalog No. ICN10139705 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10139705 5 g
ICN10139725 25 g
ICN10139780 100 g
ICN10139790 500 g
ICN10139791 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN10139705 Supplier MP Biomedicals, Inc Supplier No. 0210139705
Add to Cart
Add to Cart

Chemical Identifiers

CAS 372-75-8
Molecular Formula C6H13N3O3
Molecular Weight (g/mol) 175.19
MDL Number MFCD00064397
InChI Key RHGKLRLOHDJJDR-UHFFFAOYNA-N
Synonym l-citrulline, citrulline, h-cit-oh, delta-ureidonorvaline, sitrulline, n5-carbamoyl-l-ornithine, l-cytrulline, n5-aminocarbonyl ornithine, n delta-carbamylornithine, citrulline, l
PubChem CID 9750
ChEBI CHEBI:16349
SMILES NC(CCCNC(N)=O)C(O)=O

Specifications

Color White
Assay Percent Range 99.3%
IUPAC Name 2-amino-5-(carbamoylamino)pentanoic acid
Quantity 5 g
Formula Weight 175.19
Physical Form Crystalline Powder
Chemical Name or Material L-Citrulline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.