Learn More
Medchemexpress LLC L-Citrulline-d4 | 1217474-00-4 | 179.21 | 1 MG
SDP
Catalog No. 5000389971
Encompass_Preferred
L-Citrulline-d4 is a deuterium labeled form of L-Citrulline, an amino acid derived from ornithine in various metabolic pathways. It serves as a tracer and an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS. Deuteration, the incorporation of stable heavy isotopes into drug molecules, is used for quantitation during drug development and can influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer for quantitative analysis.
  • Functions as an internal standard for analytical methods.
  • Suitable for NMR, GC-MS, or LC-MS applications.
  • Can affect pharmacokinetic and metabolic profiles.
Catalog No. 50-003-89971 Supplier Medchemexpress LLC Supplier No. HYN0391S41MG
L-Citrulline-d4 is a deuterium labeled form of L-Citrulline, an amino acid derived from ornithine in various metabolic pathways. It serves as a tracer and an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS. Deuteration, the incorporation of stable heavy isotopes into drug molecules, is used for quantitation during drug development and can influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer for quantitative analysis.
  • Functions as an internal standard for analytical methods.
  • Suitable for NMR, GC-MS, or LC-MS applications.
  • Can affect pharmacokinetic and metabolic profiles.