Learn More
Medchemexpress LLC L-Citrulline-d7 | 2483831-24-7 | 182.23 | 5 MG
SDP
Catalog No. 5000389399
Encompass_Preferred
L-Citrulline-d7 is the deuterium-labeled form of L-Citrulline. L-Citrulline is an amino acid that originates from ornithine during the catabolism of proline or glutamine and glutamate, or from L-arginine through the arginine-citrulline pathway. This product is intended for research use only and is not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Purity: 99.83%.
Catalog No. 50-003-89399 Supplier Medchemexpress LLC Supplier No. HYN0391S55MG
L-Citrulline-d7 is the deuterium-labeled form of L-Citrulline. L-Citrulline is an amino acid that originates from ornithine during the catabolism of proline or glutamine and glutamate, or from L-arginine through the arginine-citrulline pathway. This product is intended for research use only and is not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Purity: 99.83%.