Learn More
Cerilliant Corporation Cocaine, 50-36-2, MFCD00056906, 1mL
Catalog No. NC9000749
Encompass
Chemical Formula: C17H21NO4, Molecular Weight: 303.35, 1.0 mg/mL in Acetonitrile; A Certified Spiking Solution suitable for use in LC/MS or GC/MS testing applications for forensic analysis, urine drug testing, or clinical toxicology.
Catalog No. NC9000749 Supplier Cerilliant Corporation Supplier No. C008
Chemical Formula: C17H21NO4, Molecular Weight: 303.35, 1.0 mg/mL in Acetonitrile; A Certified Spiking Solution suitable for use in LC/MS or GC/MS testing applications for forensic analysis, urine drug testing, or clinical toxicology.