Learn More
Cole-Parmer Diba Omnifit Standard Connector for 1 to 4 mm OD Tubing, 2-way, elbow
Catalog No. NC1855651 Shop All Cole-Parmer Products
Encompass
Quickly and easily connect semi-rigid tubing such as PTFE, ETFE, and FEP as well as rigid materials such as glass, PEEK, and stainless steel. Use the interchangeable components to create chemically inert fluid pathway connections and flat-bottom ports (using Gripper fittings). Fittings are autoclavable (remove caps first). Order colored caps quickly and easily color code multiple fluid lines.
Catalog No. NC1855651 Supplier Cole-Parmer Supplier No. 2193909
Quickly and easily connect semi-rigid tubing such as PTFE, ETFE, and FEP as well as rigid materials such as glass, PEEK, and stainless steel. Use the interchangeable components to create chemically inert fluid pathway connections and flat-bottom ports (using Gripper fittings). Fittings are autoclavable (remove caps first). Order colored caps quickly and easily color code multiple fluid lines.