Learn More
Medchemexpress LLC L-Cyclohexylalanine | 27527-05-5 | 171.24 | 10 G
SDP
Catalog No. 5000445913
Encompass_Preferred
L-Cyclohexylalanine is an amino acid derivative. It modifies an atrial natriuretic peptide, regulating body fluid homeostasis, blood pressure homeostasis, and vasodilation activity. It is intended for research use only.
  • Amino acid derivative
  • Modifies an atrial natriuretic peptide
  • Regulates body fluid homeostasis
  • Regulates blood pressure homeostasis
  • Exhibits vasodilation activity
  • Intended for research use only
Catalog No. 50-004-45913 Supplier Medchemexpress LLC Supplier No. HYW00245010G
L-Cyclohexylalanine is an amino acid derivative. It modifies an atrial natriuretic peptide, regulating body fluid homeostasis, blood pressure homeostasis, and vasodilation activity. It is intended for research use only.
  • Amino acid derivative
  • Modifies an atrial natriuretic peptide
  • Regulates body fluid homeostasis
  • Regulates blood pressure homeostasis
  • Exhibits vasodilation activity
  • Intended for research use only