Learn More
Medchemexpress LLC L-Cycloserine | 339-72-0 | 98.84% | 5 MG
SDP
Catalog No. 5000412559
Encompass_Preferred
L-Cycloserine is an oral inhibitor of the enzyme gamma-aminobutyric acid (GABA) transaminase (GABA-t) and branched-chain transaminases in Mycobacterium tuberculosis. It has anticonvulsant properties and inhibits the synthesis of neurotensin in mouse brains.
  • Oral inhibitor of GABA transaminase and branched-chain transaminases in Mycobacterium tuberculosis.
  • Possesses anticonvulsant properties.
  • Inhibits the synthesis of neurotensin in mouse brains.
  • Inhibits the growth of M. tuberculosis with an MIC of 0.3 μg/mL.
  • Has anticonvulsant effects in mice.
  • Enhances the inhibitory effect on GABA-T activity and boosts anticonvulsant effects in mice.
  • Reduces brain content of gangliosides.
  • Affects the level of inhibition of 3-keto-dihydrosphingosine synthase.
Catalog No. 50-004-12559 Supplier Medchemexpress LLC Supplier No. HYB11225MG
L-Cycloserine is an oral inhibitor of the enzyme gamma-aminobutyric acid (GABA) transaminase (GABA-t) and branched-chain transaminases in Mycobacterium tuberculosis. It has anticonvulsant properties and inhibits the synthesis of neurotensin in mouse brains.
  • Oral inhibitor of GABA transaminase and branched-chain transaminases in Mycobacterium tuberculosis.
  • Possesses anticonvulsant properties.
  • Inhibits the synthesis of neurotensin in mouse brains.
  • Inhibits the growth of M. tuberculosis with an MIC of 0.3 μg/mL.
  • Has anticonvulsant effects in mice.
  • Enhances the inhibitory effect on GABA-T activity and boosts anticonvulsant effects in mice.
  • Reduces brain content of gangliosides.
  • Affects the level of inhibition of 3-keto-dihydrosphingosine synthase.