Learn More
Cayman Chemical L-(+)-Cystathionine, Purity: ≥98%, CAS: 56-88-2, 10 mg
SDP
Catalog No. 502969504
Encompass_Preferred
A precursor of cysteine formed in the methionine transsulfuration pathway; deficiency can indicate the presence of metabolic disorders such as cystathioninuria
Catalog No. 50-296-9504 Supplier Cayman Chemical Supplier No. 1606110 MG
A precursor of cysteine formed in the methionine transsulfuration pathway; deficiency can indicate the presence of metabolic disorders such as cystathioninuria