Learn More
Medchemexpress LLC L-Cystathionine dihydrochloride | 61125-50-6 | 295.18 | 10 MG
SDP
Catalog No. 5000453344
Encompass_Preferred
L-Cystathionine dihydrochloride is a nonprotein thioether and a key amino acid associated with the metabolic state of sulfur-containing amino acids. It protects against Homocysteine-induced mitochondria-dependent apoptosis of vascular endothelial cells (HUVECs) and plays an important role in cardiovascular protection.
  • Is a nonprotein thioether
  • Key amino acid linked to the metabolic state of sulfur-containing amino acids
  • Protects against Homocysteine-induced mitochondria-dependent apoptosis of vascular endothelial cells (HUVECs)
  • Plays an important role in cardiovascular protection
Catalog No. 50-004-53344 Supplier Medchemexpress LLC Supplier No. HYW009749C10MG
L-Cystathionine dihydrochloride is a nonprotein thioether and a key amino acid associated with the metabolic state of sulfur-containing amino acids. It protects against Homocysteine-induced mitochondria-dependent apoptosis of vascular endothelial cells (HUVECs) and plays an important role in cardiovascular protection.
  • Is a nonprotein thioether
  • Key amino acid linked to the metabolic state of sulfur-containing amino acids
  • Protects against Homocysteine-induced mitochondria-dependent apoptosis of vascular endothelial cells (HUVECs)
  • Plays an important role in cardiovascular protection