Learn More
Medchemexpress LLC L-Cystathionine dihydrochloride | 61125-50-6 | 295.18 | 5 MG
SDP
Catalog No. 5000453136
Encompass_Preferred
L-Cystathionine dihydrochloride is a nonprotein thioether and a key amino acid involved in the metabolic state of sulfur-containing amino acids. It has been shown to protect against Homocysteine-induced mitochondria-dependent apoptosis of vascular endothelial cells (HUVECs). This suggests an important role for L-Cystathionine in cardiovascular protection.
  • Key amino acid associated with the metabolic state of sulfur-containing amino acids
  • Protects against Homocysteine-induced mitochondria-dependent apoptosis of vascular endothelial cells (HUVECs)
  • Plays an important role in cardiovascular protection
Catalog No. 50-004-53136 Supplier Medchemexpress LLC Supplier No. HYW009749C5MG
L-Cystathionine dihydrochloride is a nonprotein thioether and a key amino acid involved in the metabolic state of sulfur-containing amino acids. It has been shown to protect against Homocysteine-induced mitochondria-dependent apoptosis of vascular endothelial cells (HUVECs). This suggests an important role for L-Cystathionine in cardiovascular protection.
  • Key amino acid associated with the metabolic state of sulfur-containing amino acids
  • Protects against Homocysteine-induced mitochondria-dependent apoptosis of vascular endothelial cells (HUVECs)
  • Plays an important role in cardiovascular protection