Learn More
Medchemexpress LLC L-Cystathionine dihydrochloride | 61125-50-6 | MFCD00136759 | 99.8% | 295.18 | 100 MG
SDP
Catalog No. 5000366943
Encompass_Preferred
L-Cystathionine dihydrochloride is a nonprotein thioether and a key amino acid associated with the metabolic state of sulfur-containing amino acids. It protects against Homocysteine-induced mitochondria-dependent apoptosis of vascular endothelial cells (HUVECs) and plays an important role in cardiovascular protection.
  • Nonprotein thioether
  • Key amino acid associated with the metabolic state of sulfur-containing amino acids
  • Protects against homocysteine-induced mitochondria-dependent apoptosis of vascular endothelial cells (HUVECs)
  • Plays an important role in cardiovascular protection
Catalog No. 50-003-66943 Supplier Medchemexpress LLC Supplier No. HYW009749C100MG
L-Cystathionine dihydrochloride is a nonprotein thioether and a key amino acid associated with the metabolic state of sulfur-containing amino acids. It protects against Homocysteine-induced mitochondria-dependent apoptosis of vascular endothelial cells (HUVECs) and plays an important role in cardiovascular protection.
  • Nonprotein thioether
  • Key amino acid associated with the metabolic state of sulfur-containing amino acids
  • Protects against homocysteine-induced mitochondria-dependent apoptosis of vascular endothelial cells (HUVECs)
  • Plays an important role in cardiovascular protection