Learn More
Medchemexpress LLC L-Cystathionine dihydrochloride (Standard) | 61125-50-6 | 99.9% | 295.18 | 10 MG
SDP
Catalog No. 5000366612
Encompass_Preferred
L-Cystathionine dihydrochloride (Standard) is an analytical standard that is also a nonprotein thioether and a key amino acid linked to the metabolic state of sulfur-containing amino acids. It offers protection against Homocysteine-induced mitochondria-dependent apoptosis in vascular endothelial cells (HUVECs), indicating an important role in cardiovascular protection. This product is intended for research and analytical applications.
  • Analytical standard for L-Cystathionine dihydrochloride.
  • Used for research and analytical applications.
  • Nonprotein thioether.
  • Key amino acid in sulfur-containing amino acid metabolism.
  • Protects against Homocysteine-induced mitochondria-dependent apoptosis of HUVECs.
  • Plays a significant role in cardiovascular protection.
  • Suitable for qualitative, quantitative, and methodological research experiments using techniques like HPLC, GC, and MS.
Catalog No. 50-003-66612 Supplier Medchemexpress LLC Supplier No. HYW009749CR10MG
L-Cystathionine dihydrochloride (Standard) is an analytical standard that is also a nonprotein thioether and a key amino acid linked to the metabolic state of sulfur-containing amino acids. It offers protection against Homocysteine-induced mitochondria-dependent apoptosis in vascular endothelial cells (HUVECs), indicating an important role in cardiovascular protection. This product is intended for research and analytical applications.
  • Analytical standard for L-Cystathionine dihydrochloride.
  • Used for research and analytical applications.
  • Nonprotein thioether.
  • Key amino acid in sulfur-containing amino acid metabolism.
  • Protects against Homocysteine-induced mitochondria-dependent apoptosis of HUVECs.
  • Plays a significant role in cardiovascular protection.
  • Suitable for qualitative, quantitative, and methodological research experiments using techniques like HPLC, GC, and MS.