Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-cysteic acid (monohydrate) | 23537-25-9 | MFCD00149544 | 99.9% | 187.17 g/mol | C3H9NO6S | 500MG
SDP

Catalog No. 5000235051
Encompass_Preferred

L-Cysteic acid monohydrate is the oxidized derivative of L-cystine supplied as a monohydrate solid for use as a biochemical reagent and reference standard. It is used in enzymatic and metabolic studies and is reported to act as a substrate for GADCase/CADCaseII.

  • Oxidation product of L-cystine.
  • Reported substrate of GADCase/CADCaseII.
  • Molecular weight 187.17 g/mol.
  • Chemical formula C3H9NO6S.
  • Typical pack sizes include 500 mg, 1 g, 5 g, and 10 g.
  • High purity (99.9% as reported by supplier).

Catalog No. 50-002-35051 Supplier Medchemexpress LLC Supplier No. HY124009A500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Cysteic acid monohydrate is the oxidized derivative of L-cystine supplied as a monohydrate solid for use as a biochemical reagent and reference standard. It is used in enzymatic and metabolic studies and is reported to act as a substrate for GADCase/CADCaseII.

  • Oxidation product of L-cystine.
  • Reported substrate of GADCase/CADCaseII.
  • Molecular weight 187.17 g/mol.
  • Chemical formula C3H9NO6S.
  • Typical pack sizes include 500 mg, 1 g, 5 g, and 10 g.
  • High purity (99.9% as reported by supplier).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.