Learn More
Cayman Chemical L-Cysteine-15N-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1795787-05-1, 5 mg
SDP
Catalog No. 502912726
Encompass_Preferred
An internal standard for the quantification of L-cysteine by GC- or LC-MS
Catalog No. 50-291-2726 Supplier Cayman Chemical Supplier No. 348385 MG
An internal standard for the quantification of L-cysteine by GC- or LC-MS