Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich L-Cysteine

Catalog No. 16814925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-814-925G 25 g
16-814-925KG 2.5 kg
16-814-9100G 100 g
16-814-9500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 16-814-925G Supplier Sigma Aldrich Supplier No. 16814925G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

L-Cysteine

Specifications

CAS 52-90-4
Melting Point 240°C
Molecular Formula C3H7NO2S
Linear Formula HSCH2CH(NH2)CO2H
MDL Number MFCD00064306
Quantity 25 g
Synonym (R)-2-Amino-3-mercaptopropionic acid
Molecular Weight (g/mol) 121.16 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.