Learn More
Bioworld L-Cysteine, 1 kg
Catalog No. 50255686
Encompass_Preferred
A non-essential amino acidUsed in the production of amino acids.Used as processing aid for bakingSpecification: Appearance: White crystals or powderBoiling Point : 254 °CDensity : 1.334 g/cm3HS Code : 29309012IUPAC_Name: (2R)-2-Amino-3-sulfanylpropanoic acidMelting Point: 240°CRTECS Number: HA1600000
Catalog No. 50-255-686 Supplier Bioworld Supplier No. 403001361
Add to Cart
Add to Cart
A non-essential amino acidUsed in the production of amino acids.Used as processing aid for bakingSpecification: Appearance: White crystals or powderBoiling Point : 254 °CDensity : 1.334 g/cm3HS Code : 29309012IUPAC_Name: (2R)-2-Amino-3-sulfanylpropanoic acidMelting Point: 240°CRTECS Number: HA1600000