Learn More
Medchemexpress LLC L-cysteine-13C3, 15N | 202406-97-1 | MFCD01073449 | 99.9% | 125.13 | 13C3H7 15NO2S | 5mg
SDP
Catalog No. 5000268699
Encompass_Preferred
L-Cysteine-13C3 15N is the 13C- and 15N-labeled L-Cysteine L-Cysteine is a conditionally essential amino acid which acts as a precursor for biologically active molecules such as hydrogen sulphide (H2S) glutathione and taurine L-Cysteine suppresses ghrelin and reduces appetite in rodents and humans[1]
Catalog No. 50-002-68699 Supplier Medchemexpress LLC Supplier No. HYY0337S5MG
L-Cysteine-13C3 15N is the 13C- and 15N-labeled L-Cysteine L-Cysteine is a conditionally essential amino acid which acts as a precursor for biologically active molecules such as hydrogen sulphide (H2S) glutathione and taurine L-Cysteine suppresses ghrelin and reduces appetite in rodents and humans[1]