Learn More
AJINOMOTO USA INC L-Cysteine (L-Cys) 1Kg
Supplier:  AJINOMOTO USA INC 34151
Encompass
Chemical Name: (R)-2-Amino-3-mercaptopropionic Acid; Chemical Formula: C3H7NO2S; Molecular Weight: 121.1600; Nitrogen Content: 0.11560000; CAS Number: 52-90-4; Assay: 98.0 to 101.0%; Description: White crystals or crystalline powder; characteristic taste 1Kg
Catalog No. NC1372282