Learn More
STEMCELL Technologies L-Cysteine, Size: 250 g
SDP
Supplier:  STEMCELL Technologies 1001128
Encompass
Regulate cell signaling pathways and support protein synthesis with L-Cysteine a semi-essential amino acid found in high-protein foods (Yin et al.). Additionally L-cysteine has been shown to enhance papain activity (Homaei et al.).
Catalog No. 50-239-0432