Learn More
Medchemexpress LLC L-cysteine-d2 | 130633-02-2 | MFCD01073499 | 98.7% | 123.17 g/mol | C3H5D2NO2S | 5mg
SDP
Catalog No. 5000268222
Encompass_Preferred
L-Cysteine-d2 is the deuterium labeled L-Cysteine L-Cysteine is a conditionally essential amino acid which acts as a precursor for biologically active molecules such as hydrogen sulphide (H2S) glutathione and taurine L-Cysteine suppresses ghrelin and reduces appetite in rodents and humans[1]
Catalog No. 50-002-68222 Supplier Medchemexpress LLC Supplier No. HYY0337S65MG
Add to Cart
Add to Cart
L-Cysteine-d2 is the deuterium labeled L-Cysteine L-Cysteine is a conditionally essential amino acid which acts as a precursor for biologically active molecules such as hydrogen sulphide (H2S) glutathione and taurine L-Cysteine suppresses ghrelin and reduces appetite in rodents and humans[1]