Learn More
Medchemexpress LLC L-cysteine-d3 | 214782-32-8 | MFCD12546019 | 99.2% | 124.18 | C3H4D3NO2S | 10mg
SDP
Catalog No. 5000268225
Encompass_Preferred
L-Cysteine-d3 is the deuterium labeled L-Cysteine L-Cysteine is a conditionally essential amino acid which acts as a precursor for biologically active molecules such as hydrogen sulphide (H2S) glutathione and taurine L-Cysteine suppresses ghrelin and reduces appetite in rodents and humans[1]
Catalog No. 50-002-68225 Supplier Medchemexpress LLC Supplier No. HYY0337S510MG
L-Cysteine-d3 is the deuterium labeled L-Cysteine L-Cysteine is a conditionally essential amino acid which acts as a precursor for biologically active molecules such as hydrogen sulphide (H2S) glutathione and taurine L-Cysteine suppresses ghrelin and reduces appetite in rodents and humans[1]