Learn More
Medchemexpress LLC HY-Y0337A 1g Medchemexpress, L-Cysteine (hydrochloride) CAS:52-89-1 Purity:>98%
SDP
Catalog No. 501934686
Encompass_Preferred
Medchemexpress, HY-Y0337A 1g L-Cysteine (hydrochloride) CAS:52-89-1 L-Cysteine hydrochloride is a conditionally essential amino acid, which acts as a precursor for biologically active molecules such as hydrogen sulphide (H2S), glutathione and taurine. L-Cysteine hydrochloride suppresses ghrelin and reduces appetite in rodents and humans [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4686 Supplier Medchemexpress LLC Supplier No. HYY0337A1G
Medchemexpress, HY-Y0337A 1g L-Cysteine (hydrochloride) CAS:52-89-1 L-Cysteine hydrochloride is a conditionally essential amino acid, which acts as a precursor for biologically active molecules such as hydrogen sulphide (H2S), glutathione and taurine. L-Cysteine hydrochloride suppresses ghrelin and reduces appetite in rodents and humans [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.