Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Cysteine HCL Anhydrous, Ajinomoto

Catalog No. 502946111
Encompass
Change view
Click to view available options
Quantity:
10 kg
20 kg
Packaging:
Fiber Box
Fiber Drum
Poly Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
50-294-6111 20 kg Fiber Drum
50-294-6105 20 kg Fiber Box
50-294-6025 10 kg Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-294-6111 Supplier Thermo Scientific Production Chemicals and Services Supplier No. A34147
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 52-89-1
Molecular Weight (g/mol) 157.62

Specifications

Packaging Fiber Drum
Quantity 20 kg
Percent Purity 98.0 to 102.0%
Chemical Name or Material L-Cysteine Hydrochloride Anhydrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.