Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich L-Cysteine Hydrochloride Monohydrate

Catalog No. 778451100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-845-1100G 50 mL
77-845-11KG 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 77-845-1100G Supplier Sigma Aldrich Supplier No. 778451100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

This product is supplied from Wacker FERMOPURE™ material, but is sold forR and D use only, not for drug, household or other uses.L-Cysteine hydrochloride monohydrate can be used in the synthesis of protein photo-cross-linkers based on 2-aryl-5-carboxytetrazole , and Zn2+-doped non-linear optical crystal by conventional slow evaporation solution technique.NMDA glutamatergic receptor antagonist.FERMOPURE is a registered trademark of Wacker Chemie AG

Specifications

CAS 622-37-7
Molecular Formula C6H5N3
Linear Formula C6H5N3
MDL Number MFCD00040895
Quantity 50 mL
Synonym Phenyl azide solution
Molecular Weight (g/mol) 119.12 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.