Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Cysteine hydrochloride monohydrate, MP Biomedicals™

Catalog No. ICN1014461 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN1014461 100 g
ICN10144601 1 kg
ICN10144605 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN1014461 Supplier MP Biomedicals, Inc Supplier No. 02101446.1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4-6-7048 | C3H10ClNO3S | 175.63 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4-6-7048
Molecular Formula C3H10ClNO3S
Molecular Weight (g/mol) 175.63
MDL Number MFCD00065606
InChI Key QIJRTFXNRTXDIP-JIZZDEOASA-N
Synonym l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.h2o, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl
PubChem CID 23462
IUPAC Name (2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
SMILES C(C(C(=O)O)N)S.O.Cl

Specifications

Quantity 100 g
Formula Weight 175.64
Grade Cell Culture
Physical Form Crystals
Chemical Name or Material L-Cysteine Hydrochloride Monohydrate, Precursor in the food, pharmaceutical, personal care industries
Recommended Storage Store at Room Temperature (15° to 30°C), store desiccated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.