Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Cysteine Hydrochloride Monohydrate, MP Biomedicals

Catalog No. MP021014465 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021014465 500 g
MP021014463 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP021014465 Supplier MP Biomedicals, Inc Supplier No. 02101446.5
Add to Cart
Add to Cart

  • L-Cysteine hydrochloride monohydrate has been used in a protocol for the separation of dorsal root ganglion neurons (DRGs)
  • It has also been used in a study to examine its effect as an inhibitor in preventing enzymatic browning in apples

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C3H10ClNO3S
Molecular Weight (g/mol) 175.63
MDL Number MFCD00065606
InChI Key QIJRTFXNRTXDIP-JIZZDEOASA-N

Specifications

CAS 1880366
Melting Point 175°C (literature)
Linear Formula C3H7NO2S·HCl· H2O
Quantity 500 g
Synonym 2-propoxy-benzaldehyde, 2-propyloxybenzaldehyde, 2-n-propoxybenzaldehyde, propoxy benzaldehyde, acmc-20anok, pubchem3160, benzaldehyde,-2-propoxy, benzaldehyde,2-propoxy, aronis23744, 2,6-dichloro-4-nitropyridine 3-oxide
Solubility Information Freely soluble in water (50mg/mL).
SMILES [H+].O.[Cl-].N[C@@H](CS)C(O)=O
IUPAC Name hydrogen (2R)-2-amino-3-sulfanylpropanoic acid hydrate chloride
PubChem CID 249807
Formula Weight 175.64
Physical Form Crystals
Chemical Name or Material L-CYSTEINE HYDROCHLORIDE MONOHYDRATE
Show More Show Less
Recommended Storage Room Temperature, desiccate, protect from light, store under nitrogen
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.