Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Cysteine hydrochloride monohydrate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015297 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5297 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5297 Supplier MilliporeSigma Supelco Supplier No. PHR11021G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Cysteine hydrochloride monohydrate (LCHCMH) is a water-soluble salt of the non-essential amino acid, L-cysteine.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
TRUSTED_SUSTAINABILITY

Specifications

CAS 4-6-7048
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Linear Formula HSCH2 CH(NH2)COOH · HCl · H2O
MDL Number MFCD00065606
Type Certified Reference Material
Quantity 1 g
Formula Weight 175.63
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.