Learn More
Medchemexpress LLC L-Cysteine, L-γ-glutamyl-, 2,2,2-trifluoroacetate (1:1) | 283159-88-6 | 95.2% | 364.30 | 1 ML
SDP
Catalog No. 5000437037
Encompass_Preferred
Gamma-glutamylcysteine TFA (γ-Glutamylcysteine TFA) is an orally active, blood-brain barrier permeable dipeptide. It activates various signaling pathways like AMPK and Nrf2, while inhibiting others such as NF-κB and MAPKs. This compound regulates macrophage polarization, induces glutathione synthesis, improves metabolic dysfunction, and alleviates hepatic lipid droplet formation.
  • Applicable to research related to inflammatory bowel disease.
  • Applicable to research related to type 2 diabetes.
  • Applicable to research related to cadmium-induced neurotoxicity.
  • Applicable to research related to Alzheimer's disease.
  • Applicable to research related to cerebral ischemia/reperfusion injury.
  • Applicable to research related to neuropathy.
  • Applicable to research related to alcoholic liver disease.
Catalog No. 50-004-37037 Supplier Medchemexpress LLC Supplier No. HY113402A10MM/1ML
Gamma-glutamylcysteine TFA (γ-Glutamylcysteine TFA) is an orally active, blood-brain barrier permeable dipeptide. It activates various signaling pathways like AMPK and Nrf2, while inhibiting others such as NF-κB and MAPKs. This compound regulates macrophage polarization, induces glutathione synthesis, improves metabolic dysfunction, and alleviates hepatic lipid droplet formation.
  • Applicable to research related to inflammatory bowel disease.
  • Applicable to research related to type 2 diabetes.
  • Applicable to research related to cadmium-induced neurotoxicity.
  • Applicable to research related to Alzheimer's disease.
  • Applicable to research related to cerebral ischemia/reperfusion injury.
  • Applicable to research related to neuropathy.
  • Applicable to research related to alcoholic liver disease.