Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Cysteine Monohydrochloride, Monohydrate, FCC, 98-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18603738 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-738 25 g Amber Glass Bottle
18-603-739 100 g Amber Glass Bottle
18-603-740 500 g Amber Glass Bottle
18-603-741 1 kg Amber Glass Bottle
18-603-743 12 kg Poly Pail
18-603-742 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-603-738 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C147225GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

C1472, 7048-04-6, C3H7NO2S·HCl·H2O

Spectrum™ Chemical L-Cysteine Monohydrochloride, Monohydrate, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 4-6-7048
Molecular Formula C3H10ClNO3S
Molecular Weight (g/mol) 175.63
InChI Key QIJRTFXNRTXDIP-JIZZDEOASA-N
IUPAC Name hydrogen (2R)-2-amino-3-sulfanylpropanoic acid hydrate chloride
SMILES [H+].O.[Cl-].N[C@@H](CS)C(O)=O

Specifications

CAS Max % 100%
Quantity 25 g
Percent Purity 98 to 101.5%
Grade FCC
Ignition Residue 0.1%
Loss on Drying 8.0-12%
Optical Rotation +5.0° to +8.0°
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.