Learn More
Medchemexpress LLC HY-W017230 25mg Medchemexpress, L-Cysteinesulfinic acid (monohydrate) CAS:207121-48-0 Purity:>98%
SDP
Catalog No. 502083407
Encompass_Preferred
Medchemexpress, HY-W017230 25mg L-Cysteinesulfinic acid (monohydrate) CAS:207121-48-0 L-Cysteinesulfinic acid monohydrate is a potent agonist at several rat metabotropic glutamate receptors (mGluRs) with pEC50s of 3.92, 4.6, 3.9, 2.7, 4.0, and 3.94 for mGluR1, mGluR5, mGluR2, mGluR4, mGluR6, and mGluR8, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3407 Supplier Medchemexpress LLC Supplier No. HYW01723025MG
Medchemexpress, HY-W017230 25mg L-Cysteinesulfinic acid (monohydrate) CAS:207121-48-0 L-Cysteinesulfinic acid monohydrate is a potent agonist at several rat metabotropic glutamate receptors (mGluRs) with pEC50s of 3.92, 4.6, 3.9, 2.7, 4.0, and 3.94 for mGluR1, mGluR5, mGluR2, mGluR4, mGluR6, and mGluR8, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.