Learn More
Medchemexpress LLC L-Cystine-15N2 | 952407-61-3 | 242.29 | 1 MG
SDP
Catalog No. 5000389943
Encompass_Preferred
L-Cystine-15N2 is the 15N-labeled L-Cystine, an amino acid and intracellular thiol that plays a critical role in the regulation of cellular processes. It is for research use only.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes into drug molecules for quantitation during drug development.
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs through deuteration.
Catalog No. 50-003-89943 Supplier Medchemexpress LLC Supplier No. HYN0394S21MG
L-Cystine-15N2 is the 15N-labeled L-Cystine, an amino acid and intracellular thiol that plays a critical role in the regulation of cellular processes. It is for research use only.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes into drug molecules for quantitation during drug development.
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs through deuteration.