Learn More
Medchemexpress LLC L-Cystine-15N2 | 952407-61-3 | 242.29 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0394S25MG
Encompass_Preferred
L-Cystine-15N2 is the 15N-labeled L-Cystine. L-Cystine is an amino acid and intracellular thiol, which plays a critical role in the regulation of cellular processes. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable under recommended storage conditions.
  • For research use only.
Catalog No. 50-003-88934