Learn More
Medchemexpress LLC L-Cystine-3,3,3',3'-d4 | 1192736-38-1 | 244.33 | 1 MG
SDP
Catalog No. 5000389948
Encompass_Preferred
L-Cystine-d4 is the deuterium labeled L-Cystine. L-Cystine is an amino acid and intracellular thiol, which plays a critical role in the regulation of cellular processes. It is intended for research use only and not for sale to patients.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89948 Supplier Medchemexpress LLC Supplier No. HYN0394S11MG
L-Cystine-d4 is the deuterium labeled L-Cystine. L-Cystine is an amino acid and intracellular thiol, which plays a critical role in the regulation of cellular processes. It is intended for research use only and not for sale to patients.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.