Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-cystine-d4 | 1192736-38-1 | 98.0% | 244.33 | C6H8D4N2O4S2 | 10 MG
SDP

Catalog No. 5000295756
Encompass_Preferred

L-Cystine-d4 is the deuterium-labeled form of the amino acid L-cystine, provided as a high-purity stable isotope reagent for analytical and metabolic studies involving cystine metabolism and redox biology.

  • Deuterium-labeled for isotope tracing and internal standard use.
  • High purity (98.0%) suitable for analytical applications.
  • Molecular weight 244.33 g/mol; molecular formula C6H8D4N2O4S2.
  • Supplied as a 10 mg quantity for small-scale analytical workflows.
  • Applicable to metabolomics, proteomics, and NMR studies.

Catalog No. 50-002-95756 Supplier Medchemexpress LLC Supplier No. HYN0394S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Cystine-d4 is the deuterium-labeled form of the amino acid L-cystine, provided as a high-purity stable isotope reagent for analytical and metabolic studies involving cystine metabolism and redox biology.

  • Deuterium-labeled for isotope tracing and internal standard use.
  • High purity (98.0%) suitable for analytical applications.
  • Molecular weight 244.33 g/mol; molecular formula C6H8D4N2O4S2.
  • Supplied as a 10 mg quantity for small-scale analytical workflows.
  • Applicable to metabolomics, proteomics, and NMR studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.