Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Cystine hydrochloride Solution 10 mm amino acid in 0.1 M HCl, MilliporeSigma™ Supelco™

Catalog No. 111013991 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3991 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3991 Supplier MilliporeSigma Supelco Supplier No. 575795MLF
Add to Cart
Add to Cart

Specifications

CAS 34760-60-6
Concentration 10 mM amino acid in 0.1 M HCl
Linear Formula C6H12N2O4S2 · HCl
MDL Number MFCD00068399
Type Analytical Standard
Quantity 5 mL
Formula Weight 276.76
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.