Learn More
Medchemexpress LLC L-Cytidine | 26524-60-7 | 99.98% | 243.22 | 100 MG
SDP
Catalog No. 5000363697
Encompass_Preferred
L-Cytidine is an L-configurational form of Cytidine, a pyrimidine nucleoside and a component of RNA. It is known to control the glial glutamate cycle and can impact brain phospholipid metabolism, catecholamine synthesis, and mitochondrial function. This product is intended for research use only.
  • L-configurational form of Cytidine
  • Pyrimidine nucleoside
  • Component of RNA
  • Controls glial glutamate cycle
  • Impacts brain phospholipid metabolism
  • Affects catecholamine synthesis
  • Influences mitochondrial function
  • For research use only
Catalog No. 50-003-63697 Supplier Medchemexpress LLC Supplier No. HY148165100MG
L-Cytidine is an L-configurational form of Cytidine, a pyrimidine nucleoside and a component of RNA. It is known to control the glial glutamate cycle and can impact brain phospholipid metabolism, catecholamine synthesis, and mitochondrial function. This product is intended for research use only.
  • L-configurational form of Cytidine
  • Pyrimidine nucleoside
  • Component of RNA
  • Controls glial glutamate cycle
  • Impacts brain phospholipid metabolism
  • Affects catecholamine synthesis
  • Influences mitochondrial function
  • For research use only