Learn More
Medchemexpress LLC L-Cytidine | 26524-60-7 | MFCD02683605 | 99.98% | 243.22 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY148165500MG
Encompass_Preferred
L-Cytidine is an L-configurational form of Cytidine, a pyrimidine nucleoside and a component of RNA. Cytidine can control the glial glutamate cycle, affect brain phospholipid metabolism, catecholamine synthesis, and mitochondrial function.
  • L-configurational form of Cytidine
  • Component of RNA
  • Affects brain phospholipid metabolism
  • Controls glial glutamate cycle
  • Influences catecholamine synthesis and mitochondrial function
Catalog No. 50-003-63775