Learn More
Selleck Chemical LLC L-DAB HBR S2996-100mg
SDP
Catalog No. 503111659
Encompass_Preferred
L-DAB HBR (L-Dab Hydrobromide L-2 4-Diaminobutyric acid hydrobromide) is an inhibitor of GABA (Gamma-aminobutyric acid) transaminase with IC50 of 500 M L-DAB HBR exhibits antitumor activity
Catalog No. 50-311-1659 Supplier Selleck Chemical LLC Supplier No. S2996100MG
L-DAB HBR (L-Dab Hydrobromide L-2 4-Diaminobutyric acid hydrobromide) is an inhibitor of GABA (Gamma-aminobutyric acid) transaminase with IC50 of 500 M L-DAB HBR exhibits antitumor activity