Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Dehydroascorbic acid, 96%

Catalog No. AC250930010
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC250930010 1 g
AC250932500 250 mg
AC250930050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC250930010 Supplier Thermo Scientific Chemicals Supplier No. 250930010
  • $216.00
    $183.65 / Each of 1

    Save $32.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 490-83-5 | C6H6O6 | 174.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 490-83-5
Molecular Formula C6H6O6
Molecular Weight (g/mol) 174.11
InChI Key SBJKKFFYIZUCET-UHFFFAOYNA-N
IUPAC Name 5-(1,2-dihydroxyethyl)oxolane-2,3,4-trione
SMILES OCC(O)C1OC(=O)C(=O)C1=O

Specifications

Melting Point 228.0°C
Assay Percent Range 95.0 % min. (ELSD) (HPLC)
Quantity 1 g
Formula Weight 174.11
Chemical Name or Material L-Dehydroascorbic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

EINECSNumber 207-720-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.