Learn More
Cayman Chemical L-Deoxyalliin, Purity: ≥98%, CAS: 21593-77-1, 10 mg
SDP
Catalog No. 502953736
Encompass_Preferred
A water soluble organosulfur compound derived from garlic that has neuroprotective, antioxidative, and anti-amyloidogenic properties
Catalog No. 50-295-3736 Supplier Cayman Chemical Supplier No. 1401410 MG
A water soluble organosulfur compound derived from garlic that has neuroprotective, antioxidative, and anti-amyloidogenic properties