Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-dihydroorotic acid | 5988-19-2 | MFCD00085339 | 100.0% | 158.11 g/mol | C5H6N2O4 | 100 MG
SDP

Catalog No. 5000302956
Encompass_Preferred

L-dihydroorotic acid is an analytical reference standard used as a quality control and calibration material in chromatographic and mass spectrometry assays. It is intended for method development, qualitative and quantitative analysis, and biochemical research involving pyrimidine metabolism. The material is supplied as a high-purity solid suitable for laboratory use.

  • High purity (99.99%).
  • Intended for HPLC, GC, and MS method development and analysis.
  • Suitable as an analytical reference standard for qualitative and quantitative assays.
  • Provided as a 100 mg solid sample for laboratory use.
  • Molecular weight 158.11 g/mol; formula C5H6N2O4; CAS number 5988-19-2.

Catalog No. 50-003-02956 Supplier Medchemexpress LLC Supplier No. HYW015495R100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

L-dihydroorotic acid is an analytical reference standard used as a quality control and calibration material in chromatographic and mass spectrometry assays. It is intended for method development, qualitative and quantitative analysis, and biochemical research involving pyrimidine metabolism. The material is supplied as a high-purity solid suitable for laboratory use.

  • High purity (99.99%).
  • Intended for HPLC, GC, and MS method development and analysis.
  • Suitable as an analytical reference standard for qualitative and quantitative assays.
  • Provided as a 100 mg solid sample for laboratory use.
  • Molecular weight 158.11 g/mol; formula C5H6N2O4; CAS number 5988-19-2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.