Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-dihydroorotic acid | 5988-19-2 | MFCD00085339 | 99.9% | 158.11 g/mol | C5H6N2O4 | 5 G
SDP

Catalog No. 5000303296
Encompass_Preferred

L-dihydroorotic acid is an endogenous metabolite and substrate of dihydroorotate dehydrogenase (DHODH), used as a research reagent in biochemical and metabolic studies. Molecular formula C5H6N2O4; molecular weight 158.11 g/mol.

  • Endogenous metabolite and DHODH substrate.
  • High purity (99.94%) for reliable analytical results.
  • Molecular formula C5H6N2O4; molecular weight 158.11 g/mol.
  • Available in research-scale packages including 5 g.
  • Powder form with recommended cold storage for long-term stability.
  • Suitable for metabolic pathway and enzyme activity studies.

Catalog No. 50-003-03296 Supplier Medchemexpress LLC Supplier No. HYW0154955G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

L-dihydroorotic acid is an endogenous metabolite and substrate of dihydroorotate dehydrogenase (DHODH), used as a research reagent in biochemical and metabolic studies. Molecular formula C5H6N2O4; molecular weight 158.11 g/mol.

  • Endogenous metabolite and DHODH substrate.
  • High purity (99.94%) for reliable analytical results.
  • Molecular formula C5H6N2O4; molecular weight 158.11 g/mol.
  • Available in research-scale packages including 5 g.
  • Powder form with recommended cold storage for long-term stability.
  • Suitable for metabolic pathway and enzyme activity studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.